Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YWHAE Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: HHD-25 , Abnova™

Catalog No. 89336027 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-336-027 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-336-027 Supplier Abnova Corporation Supplier No. MAB19502
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

YWHAE Monoclonal antibody specifically detects YWHAE in Human, Mouse, Rat samples. It is validated for ICC, WB-Ce, IHC-P, IF, Flow Cyt.

Specifications

Antigen YWHAE
Applications Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone HHD-25
Conjugate Unconjugated
Description tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilon polypeptide
Gene YWHAE
Gene Alias 14-3-3E, FLJ45465, KCIP-1, MDCR, MDS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human YWHAE
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7531
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.