Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YWHAQ Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: HHF-25 , Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19507
Description
YWHAQ Monoclonal antibody specifically detects YWHAQ in Human, Mouse, Rat samples. It is validated for ICC, WB-Ce, IF, Flow Cyt.Specifications
| YWHAQ | |
| Monoclonal | |
| Unconjugated | |
| YWHAQ | |
| Rabbit | |
| 100 μg | |
| 10971 | |
| Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
| IgG |
| Immunocytochemistry, Western Blot, Immunofluorescence, Flow Cytometry | |
| HHF-25 | |
| tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta polypeptide | |
| 14-3-3, 1C5, HS1 | |
| A synthetic peptide corresponding to human YWHAQ | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction