Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YY1 Monoclonal Antibody (2C10F9)
GREENER_CHOICE

Catalog No. PIMA549173
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549173 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549173 Supplier Invitrogen™ Supplier No. MA549173
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Immunogen sequence is identical to the related mouse sequence. Positive Control - WB: human Caco-2 whole cell, human SW620 whole cell, human MDA-MB-453, human PC-3 whole cell, rat thymus tissue, mouse thymus tissue. IHC: human thyroid cancer tissue, human pancreatic ductal adenocarcinoma tissue, human laryngeal squamous cell carcinomas tissue, human renal clear cell carcinoma tissue. Flow: PC-3 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

YY1 is a ubiquitously distributed transcription factor belonging to the GLI-Kruppel class of zinc finger proteins. The protein is involved in repressing and activating a diverse number of promoters. YY1 may direct histone deacetylases and histone acetyltransferases to a promoter in order to activate or repress the promoter, thus implicating histone modification in the function of YY1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YY1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2C10F9
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene YY1
Gene Accession No. P25490, Q00899
Gene Alias AW488674; DELTA; delta transcription factor; fb59g10; INO80 complex subunit S; INO80S; NF-E1; NMP1; NMP-1; transcription factor Yy1; transcriptional repressor protein YY1; transcriptional repressor protein YY1a; Ucrbp; UCRBP transcription factor; UCR-motif DNA-binding protein; wu:fa16g07; wu:fb59g10; Yin and yang 1; Yin and Yang 1 protein; Yin Yang 1; YIN-YANG-1; yy1; YY-1; yy1 protein; YY1 transcription factor; YY1 transcription factor a; yy1a
Gene Symbols YY1
Host Species Mouse
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human YY1 (206-241aa EQKQVQIKTLEGEFSVTMWSSDEKKDIDHETVVEEQ).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22632, 24919, 7528
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.