Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ YY1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529138
Description
Recommended positive controls: 293T. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (80%), Chicken (84%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
YY1 is a ubiquitously distributed transcription factor belonging to the GLI-Kruppel class of zinc finger proteins. The protein is involved in repressing and activating a diverse number of promoters. YY1 may direct histone deacetylases and histone acetyltransferases to a promoter in order to activate or repress the promoter, thus implicating histone modification in the function of YY1.
Specifications
YY1 | |
Polyclonal | |
Unconjugated | |
YY1 | |
AW488674; DELTA; delta transcription factor; fb59g10; INO80 complex subunit S; INO80S; NF-E1; NMP1; NMP-1; transcription factor Yy1; transcriptional repressor protein YY1; transcriptional repressor protein YY1a; Ucrbp; UCRBP transcription factor; UCR-motif DNA-binding protein; wu:fa16g07; wu:fb59g10; Yin and yang 1; Yin and Yang 1 protein; Yin Yang 1; YIN-YANG-1; yy1; YY-1; yy1 protein; YY1 transcription factor; YY1 transcription factor a; yy1a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7528 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
0.88 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P25490 | |
YY1 | |
Recombinant protein encompassing a sequence within the center region of human YY1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction