Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YY1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588867
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588867 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588867 Supplier Invitrogen™ Supplier No. PA588867
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HPPMIALQPL VTDDPTQVHH HQEVILVQTR EEVVGGDDSD GLRAEDGFED QILIPVPAPA GGDDDYIEQT LVTVAAAGKS G Positive Samples: HeLa, Jurkat; Cellular Location: Nucleus matrix.

YY1 is a ubiquitously distributed transcription factor belonging to the GLI-Kruppel class of zinc finger proteins. The protein is involved in repressing and activating a diverse number of promoters. YY1 may direct histone deacetylases and histone acetyltransferases to a promoter in order to activate or repress the promoter, thus implicating histone modification in the function of YY1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YY1
Applications ChIP Assay, ELISA, Western Blot
Classification Polyclonal
Concentration 1.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene YY1
Gene Accession No. P25490, Q00899
Gene Alias AW488674; DELTA; delta transcription factor; fb59g10; INO80 complex subunit S; INO80S; NF-E1; NMP1; NMP-1; transcription factor Yy1; transcriptional repressor protein YY1; transcriptional repressor protein YY1a; Ucrbp; UCRBP transcription factor; UCR-motif DNA-binding protein; wu:fa16g07; wu:fb59g10; Yin and yang 1; Yin and Yang 1 protein; Yin Yang 1; YIN-YANG-1; yy1; YY-1; yy1 protein; YY1 transcription factor; YY1 transcription factor a; yy1a
Gene Symbols YY1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 80-160 of human YY1 (NP_0033941).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22632, 24919, 7528
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.