Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YY1 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993682
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3682 10 μL
50-199-3681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3682 Supplier Active Motif Supplier No. 61780
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

YY1 is a ubiquitously distributed transcription factor belonging to the GLI-Kruppel class of zinc finger proteins. The protein is involved in repressing and activating a diverse number of promoters. YY1 may direct histone deacetylases and histone acetyltransferases to a promoter in order to activate or repress the promoter, thus implicating histone modification in the function of YY1.

Specifications

Antigen YY1
Applications ChIP sequencing (ChIP-seq), Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene YY1
Gene Accession No. P25490
Gene Alias AW488674; DELTA; Delta transcription factor; INO80 complex subunit S; INO80S; NF-E1; NMP1; NMP-1; Transcriptional repressor protein YY1; Ucrbp; UCRBP transcription factor; UCR-motif DNA-binding protein; Yin and yang 1; Yin and Yang 1 protein; Yin Yang 1; YIN-YANG-1; YY1; YY-1; YY1 transcription factor
Gene Symbols YY1
Host Species Rabbit
Immunogen This antibody was raised against a recombinant protein corresponding to amino acids 249-414 of human YY1.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7528
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.