Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YY1 Rabbit anti-Human, Mouse, Rat, Clone: 6J6L1, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | YY1 |
---|---|
Clone | 6J6L1 |
Applications | ChIP Assay, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
YY1 Monoclonal antibody specifically detects YY1 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunofluorescence, ImmunoprecipitationSpecifications
YY1 | |
ChIP Assay, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
7528 | |
IgG | |
Affinity purified |
6J6L1 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DELTA, Delta transcription factor, INO80SYY-1, NF-E1INO80 complex subunit S, transcriptional repressor protein YY1, UCRBP, Yin and yang 1, Yin and Yang 1 protein, YIN-YANG-1, YY1 transcription factor | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human YY1 (P25490). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title