Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YY1 Recombinant Rabbit Monoclonal Antibody (6H6L1)

Catalog No. 703552
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-355-2 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-355-2 Supplier Invitrogen™ Supplier No. 703552
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Mouse, Pig, Bovine. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

YY1 is a ubiquitously distributed transcription factor belonging to the GLI-Kruppel class of zinc finger proteins. The protein is involved in repressing and activating a diverse number of promoters. YY1 may direct histone deacetylases and histone acetyltransferases to a promoter in order to activate or repress the promoter, thus implicating histone modification in the function of YY1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YY1
Applications ChIP Assay, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 6H6L1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene YY1
Gene Accession No. P25490
Gene Alias AW488674; DELTA; delta transcription factor; fb59g10; INO80 complex subunit S; INO80S; NF-E1; NMP1; NMP-1; transcription factor Yy1; transcriptional repressor protein YY1; transcriptional repressor protein YY1a; Ucrbp; UCRBP transcription factor; UCR-motif DNA-binding protein; wu:fa16g07; wu:fb59g10; Yin and yang 1; Yin and Yang 1 protein; Yin Yang 1; YIN-YANG-1; yy1; YY-1; yy1 protein; YY1 transcription factor; YY1 transcription factor a; yy1a
Gene Symbols YY1
Host Species Rabbit
Immunogen Protein corresponding to human YY1 [aa1-aa170].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7528
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.