Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YY1AP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598991
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598991 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598991 Supplier Invitrogen™ Supplier No. PA598991
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

YY1AP1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The encoded gene product presumably interacts with YY1 protein; however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YY1AP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene YY1AP1
Gene Accession No. Q9H869
Gene Alias HCCA1; HCCA2; Hepatocellular carcinoma susceptibility protein; hepatocellular carcinoma-associated protein 2; YAP; YY1 associated protein 1; YY1AP; YY1AP1; YY1-associated protein 1
Gene Symbols YY1AP1
Host Species Rabbit
Immunogen Recombinant Human YY1-associated protein 1 protein (335-466AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55249
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.