Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ YY2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567626
Description
YY2 Polyclonal Antibody for Western Blot
The protein encoded by this gene is a transcription factor that includes several Kruppel-like zinc fingers in its C-terminal region. It possesses both activation and repression domains, and it can therefore have both positive and negative effects on the transcription of target genes. This gene has an intronless coding region, and it appears to have arisen by retrotransposition of the related YY1 transcription factor gene, which is located on chromosome 14.
Specifications
YY2 | |
Polyclonal | |
Unconjugated | |
YY2 | |
transcription factor yin yang 2; transcription factor YY2; Yin and yang 2; YY2; YY-2; YY2 transcription factor; zinc finger protein 631; ZNF631 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100073351, 404281 | |
-20°C | |
Liquid |
Western Blot | |
2.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O15391, Q3TTC2 | |
YY2 | |
Synthesized peptide derived from human YY2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction