Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YY2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120469
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120469 Supplier Invitrogen™ Supplier No. PA5120469
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The target is usually found in the following locations: Nucleus. Immunogen sequence: GEGQAENPPD YSEYLKGKKL PPGGLPGIDL SDPKQLAEFT KVKPKRSKGE PPKTVPCSYS GCEKMFRDYA AMRKHLHIHG PRVHVCAECG KAFLESSKLR R.

The protein encoded by this gene is a transcription factor that includes several Kruppel-like zinc fingers in its C-terminal region. It possesses both activation and repression domains, and it can therefore have both positive and negative effects on the transcription of target genes. This gene has an intronless coding region, and it appears to have arisen by retrotransposition of the related YY1 transcription factor gene, which is located on chromosome 14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YY2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene YY2
Gene Accession No. O15391, P0C6P6, Q3TTC2
Gene Alias transcription factor yin yang 2; transcription factor YY2; Yin and yang 2; YY2; YY-2; YY2 transcription factor; zinc finger protein 631; ZNF631
Gene Symbols YY2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human YY2 (NP_996806.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100073351, 103690827, 404281
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.