Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (Z)-4-Hydroxytamoxifen
GREENER_CHOICESDP

Catalog No. 341210 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-1210 10 mg
34-125-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 34-1210 Supplier Tocris Bioscience™ Supplier No. 3412/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Metabolite of tamoxifen (Cat. No. 0999)

(Z)-4-Hydroxytamoxifen is an estrogen receptor antagonist (IC50 = 27 μM). (Z)-4-Hydroxytamoxifen is a Tamoxifen (Cat. No. 0999) metabolite; that exhibits greater potency than the parent compound. (Z)-4-Hydroxytamoxifen Also activates intein-linked inactive Cas9, reducing off-target CRISPR-mediated gene editing; system has ∽25-fold higher specificity than wtCas9. (Z)-4-Hydroxytamoxifen binds to voltage-gated sodium channels, near the intracellular gate and inhibits the sodium current by delaying channel recovery from the inactivated state (IC50 values are 297 nM and 2.1 μM for NavM and human Nav, respectively in HEK cells); this inhibition is independent of estrogen receptor modulation. Chemotherapeutic agent.

Chemical Identifiers

CAS 68047-06-3
Molecular Formula C26H29NO2

Specifications

Quantity 10 mg
Formula Weight 387.51
Purity >98%
Molecular Weight (g/mol) 387.523
Chemical Name or Material (Z)-4-Hydroxytamoxifen
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.