Learn More
MP Biomedicals™ Z-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-Fluoromethylketone
Z-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-FMK is an irreversible and cell permeable inhibitor of CPP32/Apopain.
Supplier: MP Biomedicals™ 03FK01001
Description
A member of the CIE/CED-3 family of cysteine proteases.
Specifications
White | |
1 mg | |
668 |
Store in a cool, dry place. Dessicate. | |
Z-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-Fluoromethylketone | |
Solid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov