Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Z-Guggulsterone
GREENER_CHOICE

Catalog No. 357010R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-701-0R 10 mg
35-705-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-701-0R Supplier Tocris Bioscience™ Supplier No. 3570/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Broad spectrum steroid receptor ligand. More active isomer of guggulsterone (Cat. No. 2013)

Z-Guggulsterone is a broad spectrum steroid receptor ligand; mineralocorticoid, progesterone and glucocorticoid receptor antagonist (Ki values are 37, 224 and 252 nM respectively) and weak androgen receptor agonist (Ki = 315 nM). Induces apoptosis in prostate cancer cells and inhibits angiogenesis via suppression of the VEGF-VEGFR2-Akt signaling pathway. Exhibits antilipidemic activity via antagonism of the farnesoid X receptor (FXR) and displays antiseptic, antirheumatic and anti-inflammatory activity in vivo. More active isomer of guggulsterone (Cat. No. 2013).

Specifications

Quantity 10 mg
Product Type Z-Guggulsterone
Purity >98%
Molecular Weight (g/mol) 312.45
Inhibitors Z-Guggulsterone

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.