Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (Z)-Pugnac
GREENER_CHOICESDP

Catalog No. 338410 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-841-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 33-841-0 Supplier Tocris Bioscience™ Supplier No. 3384/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

O-GlcNAcase and β-hexosaminidase inhibitor

O-GlcNAc-β-N-acetylglucosaminidase (O-GlcNAcase) and β-hexosaminidase inhibitor (Ki values are 46 and 36 nM respectively) that increases O-GlcNAc levels ∽ 2-fold in HT29 cells. Z-linked isomer is more potent than the E isomer.

Chemical Identifiers

CAS 132489-69-1
Molecular Formula C15H19N3O7

Specifications

Quantity 10 mg
Formula Weight 353.33
Purity >99%
Molecular Weight (g/mol) 353.33
Chemical Name or Material (Z)-Pugnac
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.