Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Z-Tyr-Val-Ala-Asp(OMe)-Fluoromethylketone, MP Biomedicals™

Catalog No. MP03FK01305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
3 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP03FK01305 5 mg
MP03FK01301 1 mg
MP03FK01303 3 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP03FK01305 Supplier MP Biomedicals™ Supplier No. 03FK01305

Irreversibly inhibits activity of Caspase-1 and -4. Z-Tyr-Val-Ala-Asp(OMe)-Fluoromethylketone, MP Biomedicals peptides are potent, cell permeable, irreversible inhibitors that are used for both in vitro and in vivo applications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Quantity 5 mg
Molecular Weight (g/mol) 630
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.