Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Z-Val-Ala-Asp (OMe)-FMK Inhibitor
SDP

For use in research applications

$438.00 - $1280.00

Specifications

Color White
Content And Storage Store at -20°C. Avoid freeze-thaw cycles
Product Type Z-Val-Ala-Asp (OMe)-FMK Inhibitor
Molecular Weight (g/mol) 467
Inhibitors Z-Val-Ala-Asp (OMe)-FMK
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP229392
SDP
View Documents
Novus Biologicals™
NBP229392
1 mg
Each for $438.00
Only null left
Add to Cart
 
NBP2293925
SDP
View Documents
Novus Biologicals™
NBP2293925MG
5 mg
Each for $1,280.00
Only null left
Add to Cart
 
Description

Description

Specifications

Specifications

White
Z-Val-Ala-Asp (OMe)-FMK Inhibitor
Z-Val-Ala-Asp (OMe)-FMK
Store at -20°C. Avoid freeze-thaw cycles
467
Solid
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.