Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZA20D3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982977
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2977 Supplier Bioss Supplier No. BS13560R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZA20D3 is a zinc-finger protein located on chromosome 15.

Specifications

Antigen ZA20D3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZFAND6
Gene Accession No. Q6FIF0
Gene Alias 3110005P07Rik; AN1-type zinc finger protein 6; Associated with PRK1 protein; associated with Prkcl1; Awp1; BOS_20174; HT032; LOC100349203; protein associated with PRK1; ZA20D3; ZFAND5B; ZFAND6; zgc:101121; Zinc finger A20 domain-containing protein 3; zinc finger AN1-type containing 6; zinc finger, A20 domain containing 3; zinc finger, AN1-type domain 6; zinc finger, AN1-type domain 6-like
Gene Symbols ZFAND6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZA20D3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54469
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.