Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZACN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114309
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114309 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114309 Supplier Invitrogen™ Supplier No. PA5114309
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZACN Polyclonal Antibody for ICC/IF, IHC (P), Western Blot

ZACN is a zinc-activated ligand-gated ion channel that defines a new subgroup of the cysteine-loop superfamily of ligand-gated ion channels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZACN
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene ZACN
Gene Accession No. Q401N2
Gene Alias L2; LGICZ; LGICZ1; ligand-gated ion channel subunit; ligand-gated ion channel zinc-activated 1; ligand-gated ion channel, zinc activated 1; ligand-gated ion-channel receptor L2; ZAC; ZAC1; ZACN; zinc activated ion channel; zinc activated ligand-gated ion channel 1; zinc-activated ligand-gated ion channel
Gene Symbols ZACN
Host Species Rabbit
Immunogen Synthetic peptide within Human ZAC aa 139-188.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353174
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.