Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Zacopride hydrochloride
GREENER_CHOICESDP

Catalog No. 179510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-951-0 10 mg
17-955-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-951-0 Supplier Tocris Bioscience™ Supplier No. 1795/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly potent 5-HT3 antagonist; also 5-HT4 agonist

Highly potent 5-HT3 receptor antagonist (Ki = 0.38 nM) and 5-HT4 receptor agonist (Ki = 373 nM). Antiemetic and anxiolytic following systemic administration in vivo.

Chemical Identifiers

CAS 101303-98-4
Molecular Formula C15H21Cl2N3O2

Specifications

Quantity 10 mg
Formula Weight 346.26
Purity >99%
Molecular Weight (g/mol) 346.252
Chemical Name or Material Zacopride hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.