Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZADH1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306247APC
Description
ZADH1 Polyclonal antibody specifically detects ZADH1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ZADH1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
DKFZp686P10120, EC 1.3.1.48, FLJ39091, FLJ99229,15-oxoprostaglandin 13-reductase, PGR2, PRG-2, prostaglandin reductase 2, ZADH115-oxoprostaglandin-delta13-reductase, zinc binding alcohol dehydrogenase domain containing 1, zinc binding alcohol dehydrogenase, domain containing 1, Zinc-binding alcohol dehydrogenase domain-containing protein 1 | |
Produced in rabbits immunized with E. coli-derived Human ZADH1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
145482 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction