Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZADH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554418
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554418 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554418 Supplier Invitrogen™ Supplier No. PA554418
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLSPNFREAV TLSRDCPVPL PGDGDLLVRN RFVGVNASDI NYSAGRYDPS VKPPFDIGFE GIGEVVALGL SASARYTVGQ AVAYMAPGSF AEYTVVPASI ATPVPS Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.

ZADH2 (zinc binding alcohol dehydrogenase domain containing 2) is a 377 amino acid protein that belongs to the zinc-containing alcohol dehydrogenase family and is encoded by a gene which maps to human chromosome 18. Chromosome 18 houses over 300 protein-coding genes and contains nearly 76 million bases. There are a variety of diseases associated with defects in chromosome 18-localized genes, some of which include Trisomy 18 (also known as Edwards syndrome), Niemann-Pick disease, hereditary hemorrhagic telangiectasia, erythropoietic protoporphyria and follicular lymphomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZADH2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Zadh2
Gene Accession No. Q8N4Q0
Gene Alias 15-oxoprostaglandin 13-reductase; C530046K17Rik; PRG-3; Prostaglandin reductase 3; Prostaglandin reductase-3; PTGR3; PTGR-3; Pthr3; Zadh2; zinc binding alcohol dehydrogenase domain containing 2; zinc binding alcohol dehydrogenase, domain containing 2; zinc-binding alcohol dehydrogenase domain-containing protein 2
Gene Symbols Zadh2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZADH2. Recombinant protein control fragment (Product #RP-93158).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284273
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.