Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZAG Antibody (Zagy-1), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280083AF700
This item is not returnable.
View return policy
Description
ZAG Monoclonal antibody specifically detects ZAG in Human samples. It is validated for Western Blot, ELISASpecifications
ZAG | |
Monoclonal | |
Alexa Fluor 700 | |
alpha-2-glycoprotein 1, zinc, alpha-2-glycoprotein 1, zinc-binding, ZA2G, ZAGzn-alpha-2-GP, zinc, zinc-alpha-2-glycoprotein, zn-alpha-2-glycoprotein, ZNGP1 | |
Recombinant human ZAG. | |
0.1 mL | |
Endocrinology, Immunology, Innate Immunity, Signal Transduction | |
563 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, ELISA | |
Zagy-1 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction