Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZAG Rabbit anti-Human, Rat, Clone: 7I8N3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
| Antigen | ZAG |
|---|---|
| Clone | 7I8N3 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
Description
ZAG Monoclonal antibody specifically detects ZAG in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ZAG | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| alpha-2-glycoprotein 1, zinc, alpha-2-glycoprotein 1, zinc-binding, ZA2G, ZAGzn-alpha-2-GP, zinc, zinc-alpha-2-glycoprotein, zn-alpha-2-glycoprotein, ZNGP1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ZAG (P25311). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 7I8N3 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Immunology, Innate Immunity, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 563 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title