Learn More
Promega ZAK Kinase Enzyme System, 10microg
Catalog No. PRV4244 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4244 10 μg
1 options
Catalog No. PRV4244 Supplier Promega Supplier No. V4244
Recombinant full-length human ZAK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. ZAK is a member of the MAPKKK family of signal transduction molecules and mediates gamma radiation signaling leading to cell cycle arrest.
  • Easily Screen and Profile ZAK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C