Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZAN Monoclonal Antibody (2B6)
GREENER_CHOICE

Catalog No. PIMA524422
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524422 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524422 Supplier Invitrogen™ Supplier No. MA524422
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: KEKPPDQKLV VRSSRDNYVL TQCDFEDDAK PLCDWSQVSA DDEDWVRASG PSPTGSTGAP GGYPNGEGSY LHMESNSFHR GGVARLLSPD LWEQG.

This gene encodes a sperm membrane protein that binds the zona pellucida of the egg in a species-specific manner. The encoded protein may be involved in signaling or gamete recognition. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZAN
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZAN
Gene Accession No. Q9Y493
Gene Alias ZAN; Zonadhesin; zonadhesin (gene/pseudogene)
Gene Symbols ZAN
Host Species Mouse
Immunogen ZAN (NP_003377.1, 19 a.a. ~ 113 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7455
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.