Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ZAP 180013
GREENER_CHOICESDP

Catalog No. 682150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-215-0 50 mg
68-211-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 68-215-0 Supplier Tocris Bioscience™ Supplier No. 6821/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Zap70 inhibitor; inhibits interaction with ITAMs

ZAP 180013 is a zeta-chain-associated protein kinase 70 (Zap70) inhibitor (IC50 = 1.8 μM in fluorescence polarization assay). Inhibits the interaction of Zap70 SH2 domain with immunoreceptor tyrosine-based activation motif (ITAMs), within the T-cell antigen receptor (TCR) signaling pathway.

Specifications

Chemical Name or Material 5-Chloro-N-[(4-chlorophenyl)methyl]-2-(ethylsulfonyl)-N-(2-furanylmethyl)-4-pyrimidinecarboxamide
CAS 873080-25-2
Quantity 50 mg
Molecular Formula C19H17Cl2N3O4S
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.