Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAP-70 Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1911R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1911R7 7 mL Prediluted
MS1911P0 100 μL Concentrated
MS1911P1 500 μL Concentrated
MS1911P 1 mL Concentrated
MS1911R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1911R7 Supplier Epredia Supplier No. MS1911R7

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Provides accurate, reproducible results with the Epredia™ ZAP-70 Ab-1, Mouse Monoclonal Antibody.

Mutations in ZAP-70 gene results in a form of Severe Combined Immunodeficiency Syndrome (SCID) in humans. ZAP-70 expression also defines a subset of Chronic Lymphocytic Leukaemia (CLL) in patients with unmutated Ig gene and poor clinical course.

Host Species: Mouse

Clone: 2F3.2

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: GST-fusion to tandem SH2 domains of human ZAP-70 corresponding to residues 1-254

Molecular Weight: 70kDa

Positive Control: Tonsil, Jurkat cells, T- and NK-cells

Cellular Localization: Cytoplasmic (with occasional staining of macrophage lysozymes)

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Specifications

Antigen ZAP-70 Ab-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2F3.2
Conjugate Unconjugated
Host Species Mouse
Immunogen GST-fusion to tandem SH2 domains of human ZAP-70 corresponding to residues 1-254
Quantity 7 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.