Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Zap-70 Recombinant Rabbit Monoclonal Antibody (7G0U1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535801
Description
Immunogen sequence: SDVWSYGVTM WEALSYGQKP YKKMKGPEVM AFIEQGKRME CPPECPPELY ALMSDCWIYK WEDRPDFLTV EQRMRACYYS LASKVEGPPG STQKAEAACA.
ZAP-70 is a cytosolic protein tyrosine kinase that is a member of the Syk family of proteins. ZAP-70 is expressed exclusively in T-cells and natural killer cells and is required for T-cell receptor activation. Upon T cell antigen receptor (TCR) engagement, ZAP70 is phosphorylated on tyrosines 292, 315 and 319 in the interdomain B, located between the SH2 and kinase domains. Phosphorylation of both tyrosines 315 (a Vav binding site) and 319 (a Lck binding site) enhances ZAP70 function in mediating lymphocyte signaling, while tyrosine 292 terminates the transient activation of ZAP70 and attentuates lymphocyte signaling.
Specifications
Zap-70 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P43403, P43404 | |
ZAP70 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 520-619 of human ZAP70 (NP_0010702). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Western Blot | |
7G0U1 | |
Unconjugated | |
ZAP70 | |
70 kDa zeta-associated protein; 70 kDa zeta-chain associated protein; ADMIO2; EC 2.7.10.2; FLJ17670; FLJ17679; I79_004087; IMD48; kinase ZAP70; mrtle; mur; Selective T cell defect; Srk; STCD; STD; syk-related protein tyrosine kinase; syk-related tyrosine kinase; Tyrosine-protein kinase ZAP-70; Tyrosine-protein kinase ZAP-70;TZK; TZK; ZA70; Zap70; ZAP-70; Zeta chain associated protein kinase 70kDa; zeta chain of T cell receptor associated protein kinase; zeta chain of T cell receptor associated protein kinase 70; zeta chain of T cell receptor associated protein kinase 70kDa; zeta-chain (TCR) associated protein kinase; zeta-chain (TCR) associated protein kinase 70; zeta-chain (TCR) associated protein kinase 70kDa; zeta-chain associated protein kinase 70kDa; zeta-chain associated protein kinase, 70kD | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22637, 7535 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction