Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAP-70 Mouse anti-Human, FITC, Clone: 1E7.2, BD™

Catalog No. BDB344931 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB344931 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB344931 Supplier BD Biosciences Supplier No. 344931
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZAP-70 Monoclonal antibody specifically detects ZAP-70 in Human samples. It is validated for Flow cytometry, Intracellular Staining .
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZAP-70
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1E7.2
Concentration 12.5μg/mL
Conjugate FITC
Description Vol. Per test: 20μL
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen ZAP-70 peptide 282-307 conjugated to KLH
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 7535 
Target Species Human
Form Purified
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.