Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Zap-70 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596093
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596093 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596093 Supplier Invitrogen™ Supplier No. PA596093
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPDPAAHLPF FYGSISRAEA EEHLKLAGMA DGLFLLRQCL RSLGGYVLSL VHDVRFHHFP IERQLNGTYA IAGGKAHCGP AELCEFYSRD PDGLPCNLRK PCNRPSGLEP QPGVFDCLRD AMVRDYVRQT WKLEGEALEQ AIISQAPQVE KLIATTAHER MPWYHSSLTR EEAERKLYSG AQTDGKFLLR PRKEQGTYAL SLIYGKTVYH YLISQDKAGK YCIPEGTKFD TLWQLVEYLK LKADGLIYCL KEACPNSSAS NASGAAAPTL PAHPSTLTHP QRRIDTLNSD GYTPEPARIT; Positive Samples: SW480, U251, Mouse brain, Mouse spleen; Cellular Location: Cell membrane, Cytoplasm, Peripheral membrane protein.

ZAP-70 is a cytosolic protein tyrosine kinase that is a member of the Syk family of proteins. ZAP-70 is expressed exclusively in T-cells and natural killer cells and is required for T-cell receptor activation. Upon T cell antigen receptor (TCR) engagement, ZAP70 is phosphorylated on tyrosines 292, 315 and 319 in the interdomain B, located between the SH2 and kinase domains. Phosphorylation of both tyrosines 315 (a Vav binding site) and 319 (a Lck binding site) enhances ZAP70 function in mediating lymphocyte signaling, while tyrosine 292 terminates the transient activation of ZAP70 and attentuates lymphocyte signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zap-70
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZAP70
Gene Accession No. P43403, P43404
Gene Alias 70 kDa zeta-associated protein; 70 kDa zeta-chain associated protein; ADMIO2; EC 2.7.10.2; FLJ17670; FLJ17679; I79_004087; IMD48; kinase ZAP70; mrtle; mur; Selective T cell defect; Srk; STCD; STD; syk-related protein tyrosine kinase; syk-related tyrosine kinase; Tyrosine-protein kinase ZAP-70; Tyrosine-protein kinase ZAP-70;TZK; TZK; ZA70; Zap70; ZAP-70; Zeta chain associated protein kinase 70kDa; zeta chain of T cell receptor associated protein kinase; zeta chain of T cell receptor associated protein kinase 70; zeta chain of T cell receptor associated protein kinase 70kDa; zeta-chain (TCR) associated protein kinase; zeta-chain (TCR) associated protein kinase 70; zeta-chain (TCR) associated protein kinase 70kDa; zeta-chain associated protein kinase 70kDa; zeta-chain associated protein kinase, 70kD
Gene Symbols ZAP70
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ZAP70 (NP_0010702).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22637, 301348, 7535
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.