Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAP Mouse anti-Human, Rat, Clone: 23D1.1, MilliporeSigma™

Mouse Monoclonal Antibody

$191.34 - $481.79

Specifications

Antigen ZAP
Clone 23D1.1
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABE191025
View Documents
MilliporeSigma
MABE191025UG
25 μg
Each for $191.34
Only null left
Add to Cart
 
MABE1910100
View Documents
MilliporeSigma
MABE1910100UG
100 μg
Each for $481.79
Only null left
Add to Cart
 
Description

Description

ZAP Monoclonal antibody specifically detects ZAP in Human, Rat samples. It is validated for Western Blotting.
Specifications

Specifications

ZAP
1 mg/mL
Monoclonal
Purified
RUO
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG2b κ
Protein G purified
23D1.1
Western Blot
Unconjugated
Mouse
Human, Rat
GST-tagged recombinant fragment corresponding to the N-terminal portion of rat Zinc finger CCCH-type antiviral protein 1 (ZAP).
Primary
Stable for 1 year at 2-8°C from date of receipt.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.