Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZAP70 Antibody (ZAP70/528), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311537AF700
Description
ZAP70 Monoclonal antibody specifically detects ZAP70 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ZAP70 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
ZAP70/528 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
70 kDa zeta-associated protein, EC 2.7.10, EC 2.7.10.2, SRKFLJ17679, STDFLJ17670, Syk-related tyrosine kinase, tyrosine-protein kinase ZAP-70, TZK, ZAP-70, zeta-chain (TCR) associated protein kinase (70 kD), zeta-chain (TCR) associated protein kinase 70kDa, zeta-chain associated protein kinase, 70kD | |
Recombinant full-length human ZAP70 protein (Uniprot: P43403) | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Immunology, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
7535 | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction