Learn More
Promega ZAP70 Kinase Enzyme System, 1mg
Catalog No. PRV3812 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3812 1 mg
1 options
Catalog No. PRV3812 Supplier Promega Supplier No. V3812
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human ZAP70 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. ZAP70 is a nonreceptor protein tyrosine kinase (part of the Syk/Zap70 family) that is involved in signaling by the T-cell antigen receptor.
  • Easily Screen and Profile ZAP70 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C