Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZAP70, p Tyr319 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP118896AF647
Description
ZAP70 Polyclonal antibody specifically detects ZAP70 in Human samples. It is validated for Flow CytometrySpecifications
ZAP70 | |
Polyclonal | |
Flow Cytometry | |
70 kDa zeta-associated protein, EC 2.7.10, EC 2.7.10.2, SRKFLJ17679, STDFLJ17670, Syk-related tyrosine kinase, tyrosine-protein kinase ZAP-70, TZK, ZAP-70, zeta-chain (TCR) associated protein kinase (70 kD), zeta-chain (TCR) associated protein kinase 70kDa, zeta-chain associated protein kinase, 70kD | |
The immunogen recognized by this antibody maps to a region including tyrosine 319 (when it is phosphorylated) of human zeta-chain (TCR) associated protein kinase 70kDa using the numbering given in entry NP_001070.2 (GeneID 7535). | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Immunology, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
7535 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction