Learn More
ZAP70 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301818AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
ZAP70, a 70 kDa member of the Syk tyrosine kinase family, plays a central role in lymphocyte activation and development, and is implicated in several immune disorders. Upon T cell antigen receptor (TCR) engagement, ZAP70 is phosphorylated on tyrosines 292, 315 and 319 in the interdomain B, located between the SH2 and kinase domains. Phosphorylation of both tyrosines 315 (a Vav binding site) and 319 (a Lck binding site) enhances ZAP70 function in mediating lymphocyte signaling, while tyrosine 292 terminates the transient activation of ZAP70 and attentuates lymphocyte signaling.Specifications
| ZAP70 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| P43403 | |
| ZAP70 | |
| Between 564 and 614 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| ZAP70 | |
| zeta-chain (TCR) associated protein kinase 70kDa | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7535 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.