Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAR1/Zygote arrest protein 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982965
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2965 Supplier Bioss Supplier No. BS13549R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This maternal effect gene is oocyte-specific and encodes a protein that is thought to function in the initiation of embryogenesis. A similar protein in mouse is required for female fertility. [provided by RefSeq, Jul 2013].

Specifications

Antigen ZAR1/Zygote arrest protein 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZAR1
Gene Accession No. Q86SH2
Gene Alias Oocyte-specific maternal effect factor; Z3CXXC6; ZAR1; zinc finger, 3CxxC-type 6; zygote arrest 1; Zygote arrest protein 1
Gene Symbols ZAR1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZAR1/Zygote arrest protein 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 326340
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.