Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZAR1L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP325236
Description
ZAR1L Polyclonal antibody specifically detects ZAR1L in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
ZAR1L | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
ZAR1-like protein, ZAR2, zygote arrest 1-like | |
This antibody has been engineered to specifically recognize the recombinant protein ZAR1L using the following amino acid sequence: FVRVPYGLYQGYGSTVPLGQPGLSGHKQPDWRQNMGPPTFLARPGLLVPANAPDYCIDPYKRAQLKAILSQMNPSLSPRLCKPNTK | |
100 μL | |
Primary | |
Human | |
Purified |
Immunocytochemistry | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
646799 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction