Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZAR1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559514
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559514 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559514 Supplier Invitrogen™ Supplier No. PA559514
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRTLSSCSPW DGRDPQEPLP ACGVTSPATG RRGLIRLRRD GDEAESKALP GPAEASQPQP PSRRSGADRQ EEPGQLEESG EKDAPCPQET KSKQVPGDAA SEPLRRPNFQ F Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 30%.

This gene encodes a member of the ZAR1 family that is predominantly expressed in oocytes and early embryos. The protein may function as an RNA regulator in early embryos.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZAR1L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZAR1L
Gene Accession No. A6NP61
Gene Alias XM_359419; Z3CXXC7; ZAR1L; ZAR1-like protein; ZAR2; zinc finger, 3CxxC-type 7; zygote arrest 1-like; zygotic arrest 2
Gene Symbols ZAR1L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZAR1L. Recombinant protein control fragment (Product #RP-98011).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 646799
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.