Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAR1L Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982967
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2967 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2967 Supplier Bioss Supplier No. BS13550R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the ZAR1 family that is predominantly expressed in oocytes and early embryos. The protein may function as an RNA regulator in early embryos.

Specifications

Antigen ZAR1L
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZAR1L
Gene Accession No. A6NP61
Gene Alias XM_359419; Z3CXXC7; ZAR1L; ZAR1-like protein; ZAR2; zinc finger, 3CxxC-type 7; zygote arrest 1-like; zygotic arrest 2
Gene Symbols ZAR1L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZAR1L.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 646799
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.