Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBED1 Monoclonal Antibody (OTI2H3), TrueMAB™, OriGene
SDP

Catalog No. 502539953
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9953 30 μL
50-167-5600 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9953 Supplier OriGene Supplier No. TA505045S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZBED1 Monoclonal antibody specifically detects ZBED1 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen ZBED1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2H3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ZBED1
Gene Accession No. O96006
Gene Alias Ac-like transposable element; ALTE; BED-type zinc finger domain-containing protein 1; DNA replication-related element binding factor; DREF; dREF homolog; hDREF; KIAA0785; Putative Ac-like transposable element; TRAMP; ZBED1; Zinc finger BED domain-containing protein 1; zinc finger BED-type containing 1; zinc finger, BED-type containing 1
Host Species Mouse
Immunogen Full length human recombinant protein of human ZBED1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9189
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.