Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBED3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563231
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563231 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563231 Supplier Invitrogen™ Supplier No. PA563231
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RELQAEREAL QARLRDVSRR EGALGWAPAA PPPLKDDPEG DRDGCVITKV LL Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 40%.

Axin is a key modulator of the Wnt/beta-catenin pathway and acts as a scaffold protein in phosphorylating and degrading cytoplasmic beta-catenin. ZBED3, a member of the Zinc finger BED-type domain containing superfamily, has been recently identified as an Axin-interacting protein and has been suggested to modulate the Wnt/beta-catenin signaling pathway. RNA interference (RNAi) experiments targeting ZBED3 RNA attenuated Wnt-induced beta-catenin accumulation, lymphoid enhancer binding factor-1-dependent luciferase reporter activity, and Wnt gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBED3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBED3
Gene Accession No. Q96IU2
Gene Alias 0610037K01Rik; 2610005H11Rik; AU018975; AU024588; axin-interacting protein; ZBED3; Zinc finger BED domain-containing protein 3; zinc finger BED-type containing 3; zinc finger, BED domain containing 3; zinc finger, BED type containing 3; zinc finger, BED-type containing 3
Gene Symbols ZBED3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBED3. Recombinant protein control fragment (Product #RP-95509).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84327
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.