Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZBED5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561747
Description
Immunogen sequence: TKHAAYKDKD ISFFKQHLDS PENNKPPTPK IVNTDNESAT EASYNVSYHI ALSGEAHTIG ELLIKPCAKD VVMRMFDEQY SKKIDAVQLS NSTVARR Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 37%.
This gene is unusual in that its coding sequence is mostly derived from Charlie-like DNA transposon; however, it does not appear to be an active DNA transposon as it is not flanked by terminal inverted repeats. The encoded protein is conserved among the mammalian Laurasiatheria branch. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
ZBED5 | |
Polyclonal | |
Unconjugated | |
ZBED5 | |
Buster1; Transposon-derived Buster1 transposase-like protein; ZBED5; Zinc finger BED domain-containing protein 5; zinc finger BED-type containing 5; zinc finger, BED-type containing 5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
58486 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q49AG3 | |
ZBED5 | |
Recombinant protein corresponding to Human ZBED5. Recombinant protein control fragment (Product #RP-100576). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction