Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBED5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562284 Supplier Invitrogen™ Supplier No. PA562284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IAPLLCILSY NFNTFAILNV YSKLTMFCTT NSLPMDLLLK QGSLKQEVES FCYQIVSESN DQKVGILQSE DKQLQPSVSK KSEGELSR Highest antigen sequence identity to the following orthologs: Mouse - 21%, Rat - 26%.

This gene is unusual in that its coding sequence is mostly derived from Charlie-like DNA transposon; however, it does not appear to be an active DNA transposon as it is not flanked by terminal inverted repeats. The encoded protein is conserved among the mammalian Laurasiatheria branch. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBED5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBED5
Gene Accession No. Q49AG3
Gene Alias Buster1; Transposon-derived Buster1 transposase-like protein; ZBED5; Zinc finger BED domain-containing protein 5; zinc finger BED-type containing 5; zinc finger, BED-type containing 5
Gene Symbols ZBED5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBED5. Recombinant protein control fragment (Product #RP-100577).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58486
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.