Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBP-89Rabbit anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501990076
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0076 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0076 Supplier Rockland Immunochemicals Supplier No. 100401685
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This polyclonal antibody is specific for human ZBP-89. Reactivity with ZBP-89 from other species has not been determined.

ZNF148 is involved in transcriptional regulation. ZNF148 represses the transcription of a number of genes including gastrin, stromelysin and enolase. ZNF148 binds to the G-rich box in the enhancer region of these genes.

Specifications

Antigen ZBP-89
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 90 mg/mL
Conjugate Unconjugated
Formulation Antibody with 0.1% sodium azide; pH
Gene ZNF148
Gene Accession No. Q9UQR1
Gene Alias 2210405J08Rik; AI480666; AW045217; BERF-1; Beta enolase repressor factor 1; BFCOL1; BOS_850; CACCC box-binding protein; CLL-associated antigen KW-10; fb06h05; G-rich box-binding protein; HT-BETA; LOC100344423; pHZ-52; si:dkey-101k6.6; Transcription factor BFCOL1; Transcription factor ZBP-89; wu:fb06h05; zbp89; Zbp-89; Zfp148; zfp149; zgc:165342; zinc finger binding protein 89; zinc finger DNA binding protein 89; Zinc finger DNA-binding protein 89; Zinc finger protein 148; Znf148
Gene Symbols ZNF148
Host Species Rabbit
Immunogen Purified full length ZBP-89 recombinant protein expressed in E.coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7707
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.