Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBP1/DLM-1/DAI Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP21479325UL
Description
ZBP1/DLM-1/DAI Polyclonal specifically detects ZBP1/DLM-1/DAI in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZBP1/DLM-1/DAI | |
Polyclonal | |
Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
C20orf183, chromosome 20 open reading frame 183, DAI, DLM1, DLM-1, DLM1dJ718J7.3, DNA-dependent activator of interferon regulatory factors, DNA-dependent activator of IRFs, Tumor stroma and activated macrophage protein DLM-1, ZBP1 Z-DNA binding protein 1, Z-DNA binding protein 1, Z-DNA-binding protein 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
81030 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ZBP1 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: GHGNIITRQTVSREDGSAGPRHLPSMAPGDSSTWGTLVDPWGPQDIHMERSILRRVQLGHSNEMRLHGVPSEGPAHIPPGSPPVSATAAGPEASFEARIPSPG | |
25ul | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction