Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBP89 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501564297
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4297 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4297 Supplier Bethyl Laboratories, Inc Supplier No. A303116A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan

ZNF148 is involved in transcriptional regulation. ZNF148 represses the transcription of a number of genes including gastrin, stromelysin and enolase. ZNF148 binds to the G-rich box in the enhancer region of these genes.

Specifications

Antigen ZBP89
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ZNF148
Gene Accession No. Q61624, Q9UQR1
Gene Alias BERF-1, BFCOL1, HT-BETA, ZBP-89, ZFP148, pHZ-52, CACCC box-binding protein, CLL-associated antigen KW-10, transcription factor ZBP-89, zinc finger DNA-binding protein 89
Gene Symbols Zfp148, ZNF148
Host Species Rabbit
Immunogen Between 375 and 425
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22661, 7707
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.