Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562203
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562203 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562203 Supplier Invitrogen™ Supplier No. PA562203
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CDSCGFGFSC EKLLDEHVLT CTNRHLYQNT RSYHRIVDIR DGKDSNIKAE FGEKDSSKTF SAQTDKYRGD TSQAADDSAS TTGSRK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 79%.

ZBTB1 acts as a transcriptional repressor. Represses cAMP-responsive element (CRE)-mediated transcriptional activation. In addition, has a role in translesion DNA synthesis. Requires for UV-inducible RAD18 loading, PCNA monoubiquitination, POLH recruitment to replication factories and efficient translesion DNA synthesis. Plays a key role in the transcriptional regulation of T lymphocyte development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB1
Gene Accession No. Q9Y2K1
Gene Alias 9430077A12; AI256369; brain specific protein 2; Bsg2; C430003J21Rik; KIAA0997; Zbtb1; zinc finger and BTB domain containing 1; zinc finger and BTB domain-containing protein 1; ZNF909
Gene Symbols ZBTB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB1. Recombinant protein control fragment (Product #RP-101259).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22890
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.