Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501564633
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4633 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4633 Supplier Bethyl Laboratories, Inc Supplier No. A303242AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

ZBTB1 is a zinc finger protein that also contains BTB (BR-C, ttk and bab) domain. While little is known about this protein or the related protein ZBTB2, its expression is known to be down-regulated in human bronchial epithelial cells in response to the presence of Vanadium. Recently, expression of variant ZBTB1 protein has been associated with increased age-related macular degeneration.

Specifications

Antigen ZBTB1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ZBTB1
Gene Accession No. Q91VL9, Q9Y2K1
Gene Alias ZNF909; zinc finger and BTB domain-containing protein 1
Gene Symbols Zbtb1
Host Species Rabbit
Immunogen Between 663 and 713
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22890, 268564
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.