Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584017
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584017 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584017 Supplier Invitrogen™ Supplier No. PA584017
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGEGETVQHF PLARPKSLMQ KLQCSFQTSW LKDFPWLRYS KDTGLMSCGW CQKTPADGGS VDLPPVGHDE LSRGTRNYKK TLLLRHHVST EHKLHEAN Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 93%.

This gene encodes a nuclear-localized protein that may be induced by p53 and regulates the cell cycle by inhibiting G1 to S phase progression. The encoded protein may interact with other cell cycle regulators.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB10
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB10
Gene Accession No. Q96DT7
Gene Alias 4832414A18; AU021107; RINZF; RINZFC; ZBTB10; zinc finger and BTB domain containing 10; zinc finger and BTB domain-containing protein 10; Zinc finger protein RIN ZF; zinc finger protein RINZF
Gene Symbols ZBTB10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB10. Recombinant protein control fragment (Product #RP-106292).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65986
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.